Heating Circulator, Forte HT60-M3 Highly Dynamic, Electrical: 230V, 60Hz, 15A, Temperature range: 70 to 400 deg C, Heating power: 6 kW Heating Power, Pump port: M16 x 1 male
Catalog #: CA10790-442
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought