Bath Tank B39, Stainless steel, WxDxH: 340x580x350 mmfor use with heating immersion circulator up to +150 deg C, Integrated handles in the rim Suitable for use with CORIO C/CD heating immersion circulators, Capacity: 39l
Catalog #: CA10015-842
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought