Bath Tank B33, Stainless steel, WxDxH: 900x360x200 mmfor use with heating immersion circulator up to +150 deg C, Integrated handles in the rim Suitable for use with CORIO C/CD heating immersion circulators, Capacity: 33l
Catalog #: CA10015-840
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought