Bath Tank B27, Stainless steel, WxDxH: 370x580x250 mmfor use with heating immersion circulator up to +150 deg C, Integrated handles in the rim Suitable for use with CORIO C/CD heating immersion circulators, Capacity: 27l
Catalog #: CA10015-838
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought