Bath Tank B17, Stainless steel, WxDxH: 370x400x250 mmfor use with heating immersion circulator up to +150 deg C, Integrated handles in the rim Suitable for use with CORIO C/CD heating immersion circulators, Capacity: 17l
Catalog #: CA10015-834
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought