Bath Tank B13, Stainless steel, WxDxH: 370x400x200 mmfor use with heating immersion circulator up to +150 deg C, Integrated handles in the rim Suitable for use with CORIO C/CD heating immersion circulators, Capacity: 13l
Catalog #: CA10015-832
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought