Transparent bath tank BT5, 5 l, Model: BT5, WxDxH: 23x38x38 cm, Opening size: 15Wx15Lx15D cm, Temperature range:upto +100 deg C
Catalog #: CA10015-822
Supplier: JML SCIENCES INC CA
Transparent bath tank BT5, 5 l, Model: BT5, WxDxH: 23x38x38 cm, Opening size: 15Wx15Lx15D cm, Temperature range:upto +100 deg C
Catalog #: CA10015-822
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
73,15 $
Shimpo Compact LCD Contact and Contact/Non-Contact Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
518,81 $
VWR® THree-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
142,45 $ to 2 173,12 $