Circulator, CD-200F, Refrigerated/Heating, Pump: 15 L/min; 4.8 PSI max, Cooling capacity: 220 W (at 20 deg C), 16 0 W (at 0 deg C), 60 W (at -20 deg C), Heating capacity: 1 kW, Height: 25.6in, Length: 15.4in, Width: 9.4in, Temperature range: -20 to +150 C, Electrical: 115v, Bath capacity: 3-4l
Catalog #: CA10015-818
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought