Circulator,C-B5, Open Heating Bath, Tank material: Stainless Steel, Pump6 L/min; 1.5 psi, Temperature Range: 20 to 100C, Heating Capacity: 1 kW, feature volume graduations in L and Qt, Bath capacity: 5l
Catalog #: CA10015-808
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought