MicroPlate Rotor; Holds: 4 x standard microplate; 2 deep well plates, Material: Aluminum, For Use With (Equipment): SRF110
Catalog #: 76499-970
Supplier: Thermo Fisher Scientific
MicroPlate Rotor; Holds: 4 x standard microplate; 2 deep well plates, Material: Aluminum, For Use With (Equipment): SRF110
Catalog #: 76499-970
Supplier: Thermo Fisher Scientific
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$48.82 to $442.63
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$31.17 to $282.75
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
$73.15
Shimpo Compact LCD Contact and Contact/Non-Contact Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$518.81