Circulator,C-BT9, Open Heating Bath, Tank material: Polycarbonate, Pump6 L/min; 1.5 psi, Temperature Range: 20 to 100C, Heating Capacity: 1 kW, feature volume graduations in L and Qt, Bath capacity: 9l
Catalog #: CA10015-802
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought