Immersion Circulator, Heating capacity: 1 kW, Maximum volume: 30 L, Pump capacity: 6 L/min; 1.5 psi, Temperature range: 20 to 100C
Catalog #: CA10015-798
Supplier: JML SCIENCES INC CA
Immersion Circulator, Heating capacity: 1 kW, Maximum volume: 30 L, Pump capacity: 6 L/min; 1.5 psi, Temperature range: 20 to 100C
Catalog #: CA10015-798
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: VWR International
$73.15
Shimpo LCD Contact/Non-Contact Handheld Laser Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$771.89
Shimpo Compact LCD Contact and Contact/Non-Contact Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
$518.81