Circulator, CD-600F, Refrigerated/Heating, Pump: 15 L/min; 4.8 PSI max, Cooling capacity: 220 W (at 20 deg C), 16 0 W (at 0 deg C), 60 W (at -20 deg C), Heating capacity: 1 kW, Height: 27.2in, Length: 18.5in, Width: 16in, Temperature range: -35 to +150 C, Electrical: 115v, Bath capacity: 5-7.5l
Catalog #: CA10015-754
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought