Circulator,CD-BT5, Open Heating Bath, Tank material: Polycarbonate, Pump: 15 L/min; 5.1 psi, Temperature Range: 20 to 100C, Heating Capacity: 1 kW, features a bright LED display with easy user interface. Includes pump connections for external fluid flow Bath capacity: 5l
Catalog #: CA10015-726
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought