Circulator,C-B19, Open Heating Bath, Tank material: Stainless Steel, Pump6 L/min; 1.5 psi, Temperature Range: 20 to 100C, Heating Capacity: 1 kW, feature volume graduations in L and Qt, Bath capacity: 19l
Catalog #: CA10015-720
Supplier: JML SCIENCES INC CA
Circulator,C-B19, Open Heating Bath, Tank material: Stainless Steel, Pump6 L/min; 1.5 psi, Temperature Range: 20 to 100C, Heating Capacity: 1 kW, feature volume graduations in L and Qt, Bath capacity: 19l
Catalog #: CA10015-720
Supplier: JML SCIENCES INC CA
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
31,17 $ to 282,75 $
VWR® Four-Channel Electronic Timer and Clock with Certificate of Calibration
Supplier: VWR International
89,92 $ to 1 797,48 $
VWR® Single Channel Electronic Timer with Memory and Clock and Certificate of Calibration
Supplier: VWR International
75,03 $ to 739,18 $
Shimpo Compact LCD Contact and Contact/Non-Contact Tachometers, Nidec Shimpo America
Supplier: Nidec Shimpo America
518,81 $