ROTAVISC me-vi HELI Complete, determines the viscosity of medium viscous liquids in all areas of application ranging from the laboratory to quality control, includes a standard spindle set (SP 7-SP 12), a protective bracket, temperature sensor and HELISTAND stand incl. T-spindle set
Catalog #: 76499-920
Supplier: IKA Works
Customers Who Bought This Also Bought