ROTAVISC lo-vi HELI Complete, determines the viscosity of low-viscosity liquids in all areas of application ranging from the laboratory to quality control, includes a standard spindle set (SP 1-SP 4), a protective bracket, temperature sensor and HELISTAND stand incl. T-spindle set.
Catalog #: 76499-918
Supplier: IKA WORKS, INC. MX
Customers Who Bought This Also Bought