ROTAVISC hi-vi II HELI Complete, determines the viscosity of extremely viscous liquids in all areas of application ranging from the laboratory to quality control, includes a standard spindle set (SP 7-SP 12), a protective bracket, temperature sensor and HELISTAND stand incl. T-spindle set.
Catalog #: 76499-904
Supplier: AVANTOR PERFORMANCE MATERIALS MX
Customers Who Bought This Also Bought