Linearity FD Cardiac Markers for Ortho Vitros, assayed quality control material consisting of five levels of human serum, Format: Freeze Dried, Storage: 3 days when stored at 2-8C, Size: 5 x 1 mL
Catalog #: 76499-408
Supplier: Audit Microcontrols
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.