Beaker, DWB 600, double-walled, has a filling volume of 600 ml. It can be connected to an external cooling or heating source and is therefore suitable for tempering the sample inside the beaker.
Catalog #: 76499-096
Supplier: IKA Works
Beaker, DWB 600, double-walled, has a filling volume of 600 ml. It can be connected to an external cooling or heating source and is therefore suitable for tempering the sample inside the beaker.
Catalog #: 76499-096
Supplier: IKA Works
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
885,46 $ to 3 541,82 $
Ethanol denatured ACS (denatured with methanol and ethyl acetate)
Supplier: INNOVA CHEMICALS CA
258,00 $
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
795,94 $ to 2 497,03 $
Fluoride Standard, 1 ml = 0.1 mg F-, 100 ppm F-, Ricca Chemical Company
Supplier: Ricca Chemical
162,76 $