VOL-C-RTD-4.2 for VOLS-1,suitable for expensive and valuable samples, A temperature sensor is already integrated in the chamber and thus allows for temperature measurement during viscosity determination, required to use the VOL-SP-4.2 spindle in combination with 4.2ml chamber VOL-C-RTD-4.2
Catalog #: 76499-092
Supplier: IKA Works
Customers Who Bought This Also Bought