VOL-C-RTD-3.8 for VOLS-1 is suitable for expensive and valuable samples. A temperature sensor is integrated in the chamber and allows for temperature measurement during viscosity determination. It is required to use the VOL-SP-3.8 in combination with the VOL-C-RTD-3.8.
Catalog #: 76499-090
Supplier: IKA Works
Customers Who Bought This Also Bought