VOL-SP-13.5 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 13.5 ml.
Catalog #: 76499-080
Supplier: IKA Works
VOL-SP-13.5 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 13.5 ml.
Catalog #: 76499-080
Supplier: IKA Works
Customers Who Bought This Also Bought
Iron, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
194,37 $
Methanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®
Supplier: VWR International
260,01 $ to 668,36 $
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
316,00 $ to 5 024,40 $
Mercury(II) thiocyanate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
323,61 $