VOL-SP-11 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 11 ml
Catalog #: 76499-078
Supplier: IKA Works
VOL-SP-11 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 11 ml
Catalog #: 76499-078
Supplier: IKA Works
Customers Who Bought This Also Bought
Chloroform ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$782.44 to $2,358.46
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$170.25 to $676.42
Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
$729.52
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
$2,423.69
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
$90.42 to $94.61
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$42.99
Potassium Chloride Conductivity Standard, 1 mS/cm (1000 µmho/cm) at 25 °C, Ricca Chemical Company
Supplier: Ricca Chemical
$99.27