VOL-SP-10.4 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 10.4 ml.
Catalog #: 76499-076
Supplier: IKA Works
VOL-SP-10.4 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 10.4 ml.
Catalog #: 76499-076
Supplier: IKA Works
Customers Who Bought This Also Bought
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$599.10 to $2,396.15
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$131.08 to $405.43
Absorbant Paper Points, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$60.77 to $60.77