VOL-SP-7.1 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 7.1 ml.
Catalog #: 76499-074
Supplier: IKA Works
VOL-SP-7.1 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 7.1 ml.
Catalog #: 76499-074
Supplier: IKA Works
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Exel International Brand Quality Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
56,12 $ to 443,54 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
1 313,27 $ to 5 149,78 $
Flow cells for MinION devices, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
7 900,00 $