VOL-SP-9.4 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 9.4 ml.
Catalog #: 76499-070
Supplier: IKA Works
VOL-SP-9.4 is an optional spindle for an extended viscosity range. It is especially suitable for sample volumes of 9.4 ml.
Catalog #: 76499-070
Supplier: IKA Works
Customers Who Bought This Also Bought
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
147,27 $ to 464,44 $
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
73,48 $ to 4 408,00 $
Masterflex® L/S® Precision Variable-Speed Console Drives, Avantor®
Supplier: Avantor Fluid Handling
3 286,40 $
Acetic acid glacial ≥99.7%, ChromAR® for liquid chromatography, for TLC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
195,95 $ to 1 175,58 $