ContactUsLinkComponent
Respirator, Particulate, 9205+, N95, three-panel, flat-fold disposable respirator designed to help provide comfortable, reliable worker protection against non-oil based particles, Designed to direct exhaled air away from the nose panel, helping reduce eyewear fogging, Soft inner materials
Catalog #: CA76499-326
Supplier:  3M
Respirator, Particulate, 9205+, N95, three-panel, flat-fold disposable respirator designed to help provide comfortable, reliable worker protection against non-oil based particles, Designed to direct exhaled air away from the nose panel, helping reduce eyewear fogging, Soft inner materials
Catalog #: CA76499-326
Supplier:  3M
Customers Who Bought This Also Bought