TUBE CENTRIFUGE STERILE 50ML CS500
Catalog # CA104013-046
Supplier: DBIOMED INC. MS
TUBE CENTRIFUGE STERILE 50ML CS500
Catalog # CA104013-046
Supplier: DBIOMED INC. MS
Supplier Number: 3181-345
Frequently Bought Together
Acrodisc® Syringe Filters, 25 mm
Supplier: Cytiva
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls
Supplier: Graphic Controls
Extracta™ DNA Prep for PCR, Quantabio
Supplier: Quantabio
WypAll® X60 Wipers
Supplier: Kimberly-Clark
VWR® Multipurpose Containers, HDPE
Supplier: VWR International
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
MOUNTING MEDIUM WITH DAPI 100 TESTS
Supplier: Enzo Life Sciences
Lensx® 90 70% Rayon/30% Hemp Nonwoven Cleanroom Wipers, Berkshire
Supplier: Berkshire
Single-Use Needles, BD Medical
Supplier: BD