ContactUsLinkComponent
Pipette Tip, Low Retention Filter, Sterile, with complete protection from sample carry-over, Incorporating a patented filter that is made from high molecular weight polyethylene, filter allow the pipettor to draw by venting through a tortuous sponge-like pathway, Size: 1ML
Catalog #: CA104013-042
Supplier:  DBIOMED INC. MS
Pipette Tip, Low Retention Filter, Sterile, with complete protection from sample carry-over, Incorporating a patented filter that is made from high molecular weight polyethylene, filter allow the pipettor to draw by venting through a tortuous sponge-like pathway, Size: 1ML
Catalog #: CA104013-042
Supplier:  DBIOMED INC. MS
Customers Who Bought This Also Bought