ContactUsLinkComponent
Pipette Tip, Extended Filter, with complete protection from sample carry-over, Incorporating a patented filter that is made from high molecular weight polyethylene, filter allow the pipettor to draw by venting through a tortuous sponge-like pathway, pores have an average size of 10um, Size: 10Ul
Catalog #: CA104013-034
Supplier:  DBIOMED INC. MS
Pipette Tip, Extended Filter, with complete protection from sample carry-over, Incorporating a patented filter that is made from high molecular weight polyethylene, filter allow the pipettor to draw by venting through a tortuous sponge-like pathway, pores have an average size of 10um, Size: 10Ul
Catalog #: CA104013-034
Supplier:  DBIOMED INC. MS
Customers Who Bought This Also Bought