Centifuge, Multifuge X4R Pro TX-1000 Tc package, Rotor: TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters, Type: Benchtop, Refrigerated, Display: Bright LCD Push-Button User Interface, Microprocessor controller, Voltage: 120v
Catalog #: MP76326-220
Supplier: Thermo Fisher Scientific