Vols-1, Adapter to measure the viscosity of especially valuable and expensive samples. A double jacket with temperature sensor to quickly heat up or cool down the sample as well as a coaxial cylinder system for the analysis of the sample at a specific shear rate is included.
Catalog #: 76499-056
Supplier: IKA Works
Customers Who Bought This Also Bought