Elvas-1, extremely low viscosity adapter spindle set includes a double jacket to quickly heat up or cool down the sample as well as a coaxial cylinder system for the analysis of the sample at a specific shear rate and shear stress, viscosity is 2000 mPas.
Catalog #: 76499-052
Supplier: IKA Works
Customers Who Bought This Also Bought