DIN spindle set (O-DINS-1) measures viscosities according to DIN 53019 and can be used with customer-specific vessels, includes a single-wall-open-jacket to quickly dive into sample liquid as well as coaxial cylinder system for analysis of sample, maximum measurable viscosity is 50,000 mPas.
Catalog #: 76499-048
Supplier: IKA Works
Customers Who Bought This Also Bought