VAN-SP-5, vane spindle, allows the viscosity measurement of suspensions.
Catalog #: 76499-038
Supplier: IKA Works
VAN-SP-5, vane spindle, allows the viscosity measurement of suspensions.
Catalog #: 76499-038
Supplier: IKA Works
Customers Who Bought This Also Bought
Sol Care® Vacuum Activated Blood Collection Safety Tube Holder Set, Sol M
Supplier: SOL-MILLENNIUM MEDICAL
49,36 $ to 387,01 $
di-Sodium tetraborate, Spectromelt® B 10 for trace metal analysis, Supelco®
Supplier: MilliporeSigma
412,90 $
Rapid Response™ COVID-19 Antigen Rapid Test Cassettes - At Home, BTNX
Supplier: BTNX INC CA
21,01 $ to 5 041,08 $
Peroxidase Test Method: colorimetric with test strips MQuant™
Supplier: MilliporeSigma
306,39 $ to 11 725,11 $