Order Entry
ContactUsLinkComponent
Screw-cap microtubes, 2.0mL, amber color, with O-ring, sterile, molded graduations, amber caps attached, 100 per bag, 1000/case
Free-Standing Light-Blocking Screw-Cap Microcentrifuge Tube, 2.0 ml
Catalog #: 470335-056
Supplier:  MTC BIO, INC.
Screw-cap microtubes, 2.0mL, amber color, with O-ring, sterile, molded graduations, amber caps attached, 100 per bag, 1000/case
Free-Standing Light-Blocking Screw-Cap Microcentrifuge Tube, 2.0 ml
Catalog #: 470335-056
Supplier:  MTC BIO, INC.

Specifications

  • Volume:
    2.0 ml
  • Color:
    Amber
  • Description:
    SureSeal screw-cap tube
  • Cat. no.:
    470335-056
  • Maximum RCF:
    22000×g

Specifications

About this item

Free-standing microcentrifuge tube in light-blocking transparent amber color can hold up to 2.0 ml.

  • Made of ultra-clear polypropylene
  • Light-blocking amber color preserves integrity of sample
  • Leak-proof O-ring seal
  • RNase, DNase, and pyrogen free
  • Molded graduations

These tubes also come sterile and with caps assembled for quick and easy usage. May be centrifuged up to 22000×g.

Customers Who Bought This Also Bought