ContactUsLinkComponent
Cartridge, Nominal length: 20in, 0.1um Polyethersulphone membrane, BF/226 Bayonet end cap with external Silicone o-rings. PROPOR MR filters have been specifically designed for fast, effective and economical removal of mycoplasma from cell culture media in the biopharmaceutical industry.
Catalog #: 76498-048
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 0.1um Polyethersulphone membrane, BF/226 Bayonet end cap with external Silicone o-rings. PROPOR MR filters have been specifically designed for fast, effective and economical removal of mycoplasma from cell culture media in the biopharmaceutical industry.
Catalog #: 76498-048
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought