Mini Capsule, Double Size, with 10um Pleated Polypropylene Depth Media, 1/2in NPT (Male) connections and vent/draing valves with silicone seal, encapsulated design maximizes efficiency by providing faster, easier change-out without laborious cleaning procedures.
Catalog #: 76497-476
Supplier: PARKER HANNIFIN TE
Customers Who Bought This Also Bought