ContactUsLinkComponent
Mini Capsule, Double Size, with 2.5um Pleated Polypropylene Depth Media, 1/2in Hose barb connections and vent/draing valves with silicone seal, encapsulated design maximizes efficiency by providing faster, easier change-out without laborious cleaning procedures.
Catalog #: 76497-412
Supplier:  PARKER HANNIFIN TE
Mini Capsule, Double Size, with 2.5um Pleated Polypropylene Depth Media, 1/2in Hose barb connections and vent/draing valves with silicone seal, encapsulated design maximizes efficiency by providing faster, easier change-out without laborious cleaning procedures.
Catalog #: 76497-412
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought