ContactUsLinkComponent
Cartridge, Nominal length: 10in, 20um Nominal Pleated 316 Stainless Steel Media, DOE with EPDM o-rings, ideal solution in applications where traditional polymer based filters are limited by compatibility, exposure time or a combination of high temperature and viscosity.
Catalog #: 76497-266
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, 20um Nominal Pleated 316 Stainless Steel Media, DOE with EPDM o-rings, ideal solution in applications where traditional polymer based filters are limited by compatibility, exposure time or a combination of high temperature and viscosity.
Catalog #: 76497-266
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought