ContactUsLinkComponent
Cartridge, Nominal length: 10in, .2um Pleated PTFE membrane, SOE Fin/222 end cap with external EPDM o-rings, sterile gas filters combine proven depth filter technology and a pleated construction to provide retention down to 0.01 micron in gas.
Catalog #: 76497-144
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, .2um Pleated PTFE membrane, SOE Fin/222 end cap with external EPDM o-rings, sterile gas filters combine proven depth filter technology and a pleated construction to provide retention down to 0.01 micron in gas.
Catalog #: 76497-144
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought