ContactUsLinkComponent
Cartridge, Nominal length: 5in, .2um Pleated PTFE membrane, SOE Flat Top/222 end cap with external EPDM o-rings, sterilization filters are part of a new generation of filter sets the standard with an unrivalled combination of efficiency, flow rate and strength.
Catalog #: 76497-110
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 5in, .2um Pleated PTFE membrane, SOE Flat Top/222 end cap with external EPDM o-rings, sterilization filters are part of a new generation of filter sets the standard with an unrivalled combination of efficiency, flow rate and strength.
Catalog #: 76497-110
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought