ContactUsLinkComponent
Cartridge, Nominal length: 20in, 20um Pleated Polypropylene media, SOE Flat Top/222 end cap with external EPDM o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
Catalog #: 76497-054
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 20um Pleated Polypropylene media, SOE Flat Top/222 end cap with external EPDM o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
Catalog #: 76497-054
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought