CARTRIDGE 10IN 20UM PP FLAT TOP/222 EPDM
Catalog # 76497-040
Supplier: PARKER HANNIFIN TE
Frequently Bought Together
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$144.40 to $1,035.64
Silver nitrate 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$187.67 to $2,052.36
Sodium hydroxide 40% (w/v) in aqueous solution for Kjeldahl nitrogen analysis nitrogen-free
Supplier: Ricca Chemical
$670.90
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$73.73 to $459.25
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
$411.94 to $1,303.38
Hanus solution (Iodine bromine) for iodine absorption number of fats and oils
Supplier: Ricca Chemical
$506.58