ContactUsLinkComponent
Cartridge, Nominal length: 30in, 0.6um Pleated Polypropylene media, SOE Fin/222 end cap with external EPDM o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
Catalog #: 76496-944
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 30in, 0.6um Pleated Polypropylene media, SOE Fin/222 end cap with external EPDM o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
Catalog #: 76496-944
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought