Cartridge, Nominal length: 10in, 1.0um Pleated Polypropylene media, SOE Fin/222 end cap with external EPDM o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
Catalog #: 76496-918
Supplier: PARKER HANNIFIN TE
Customers Who Bought This Also Bought