ContactUsLinkComponent
Cartridge, Nominal length: 30in, 1.0um Pleated Polypropylene media, SOE BF/226 Bayonet end cap with external silicone o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
Catalog #: 76496-778
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 30in, 1.0um Pleated Polypropylene media, SOE BF/226 Bayonet end cap with external silicone o-rings, utilized for the clarification and prefiltration of a wide range of products in the fermentation and fine chemical industries.
Catalog #: 76496-778
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought