ContactUsLinkComponent
Cartridge, Nominal length: 5in, 5um Pleated Polypropylene Depth Media, SOE Flat Top/222 end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-588
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 5in, 5um Pleated Polypropylene Depth Media, SOE Flat Top/222 end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-588
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought