Cartridge, Nominal length: 30in, 3um Pleated Polypropylene Depth Media, SOE Fin/222t end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-516
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 30in, 3um Pleated Polypropylene Depth Media, SOE Fin/222t end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-516
Supplier: PARKER HANNIFIN TE
Customers Who Bought This Also Bought
Nalgene® Laboratory Notebooks, Thermo Scientific
Supplier: Thermo Fisher Scientific
$172.57 to $531.06
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
$1,747.88
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
$541.80
Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Supplier: Thermo Fisher Scientific
$919.35 to $933.31
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$121.35 to $993.13
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
$678.63 to $1,914.33
Chemware® Laboratory Matting, PTFE, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Life Sciences
$1,947.60